Learn More
Perkin Elmer US LLC Front Ferrule for 2.4 mm O.D. Tubing
Catalog No. 502016309
Encompass_Preferred
Requires back ferrule (B3001334), and nut (B3001335). These components are compatible with tubing B0017998 which is sold by the meter (m).
Catalog No. 50-201-6309 Supplier Perkin Elmer US LLC Supplier No. B3001333
Requires back ferrule (B3001334), and nut (B3001335). These components are compatible with tubing B0017998 which is sold by the meter (m).