Learn More
Cayman Chemical REquol Natural Products Extrac
SDP
Catalog No. 502889362
Encompass_Preferred
An agonist of ERα and active metabolite of daidzein; induces ERα-mediated gene transcription selectively over ERβ-mediated transcription in HEC-1 cells expressing the human receptors in a reporter assay (EC50s = 66 and 330 nM, respectively)
Catalog No. 50-288-9362 Supplier Cayman Chemical Supplier No. 100101721 MG
Add to Cart
Add to Cart
An agonist of ERα and active metabolite of daidzein; induces ERα-mediated gene transcription selectively over ERβ-mediated transcription in HEC-1 cells expressing the human receptors in a reporter assay (EC50s = 66 and 330 nM, respectively)