Learn More
Aviva Systems Biology Corp RERE Antibody - N-terminal region (ARP32713_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32713_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RERE. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RERE. Alternate names: ARG, ARP, ATN1L, DNB1, FLJ38775, KIAA0458ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54719