Learn More
ABclonal Technology RERE Rabbit pAb
SDP
Supplier:  ABclonal Technology A15255500UL
Encompass_Preferred
This gene encodes a member of the atrophin family of arginine-glutamic acid (RE) dipeptide repeat-containing proteins. The encoded protein co-localizes with a transcription factor in the nucleus, and its overexpression triggers apoptosis. A similar protein in mouse associates with histone deacetylase and is thought to function as a transcriptional co-repressor during embryonic development. Multiple transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-07089