Learn More
Aviva Systems Biology Corp RERG Antibody - C-terminal region (ARP59687_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59687_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RERG. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human RERG. Alternate names: MGC15754ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-286-778