Learn More
Aviva Systems Biology Corp Rerg Antibody - middle region (ARP59688_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59688_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rerg. It was validated on Western Blot. . Alternate names: RGD1562829ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-286-779