Learn More
DIGI KEY CORPORATION RES 470 OHM 1 1/8W 0805
SDP
Supplier:  DIGI KEY CORPORATION 311470CRCTND
Encompass
470 Ohms ±1% 0.125W, 1/8W Chip Resistor 0805 (2012 Metric) Moisture Resistant Thick Film
Catalog No. NC2308885