Learn More
Chemglass Life Sciences 1000mL Reservoir, Chromatography, 50/30 Lower Ball
SDP
Catalog No. CG120034
Encompass_Preferred
1000mL Reservoir, Chromatography, 50/30 Lower Ball. Reservoir for use with CG-1199 and CG-1200 columns. Reservoir has a top 35/20 socket joint for use with CG-1201 flow control adapter and the listed size lower ball joint.
Catalog No. CG120034 Supplier Chemglass Life Sciences Supplier No. CG120034
1000mL Reservoir, Chromatography, 50/30 Lower Ball. Reservoir for use with CG-1199 and CG-1200 columns. Reservoir has a top 35/20 socket joint for use with CG-1201 flow control adapter and the listed size lower ball joint.