Learn More
Agilent Technologies Agilent reservoir, 12 column, polypropylene, 21 mL/column, 252 mL maximum, pyramid base geometries, 44 mm height, 25/pk
Catalog No. 50995865
Encompass
Agilent reservoir, 12 column, polypropylene, 21 mL/column, 252 mL maximum, pyramid base geometries, 44 mm height, 25/pk
Catalog No. 50-995-865 Supplier Agilent Technologies Supplier No. 201256100
Agilent reservoir, 12 column, polypropylene, 21 mL/column, 252 mL maximum, pyramid base geometries, 44 mm height, 25/pk