Learn More
Agilent Technologies Agilent reservoir, 12 column, polypropylene, 7 mL/column, 84 mL maximum, pyramid base geometries, 19 mm height, 25/pk
Catalog No. 50995871
Encompass
Agilent reservoir, 12 column, polypropylene, 7 mL/column, 84 mL maximum, pyramid base geometries, 19 mm height, 25/pk
Catalog No. 50-995-871 Supplier Agilent Technologies Supplier No. 201280100
Agilent reservoir, 12 column, polypropylene, 7 mL/column, 84 mL maximum, pyramid base geometries, 19 mm height, 25/pk