Learn More
Agilent Technologies RES SEAHORSE 16/384 4.9ML 25CS
Catalog No. 50995875
Encompass
Seahorse 16 row polypropylene reservoir, 4.9ml/row, 54ml max, pyramid base geometries, 19mm height, case of 25
Catalog No. 50-995-875 Supplier Agilent Technologies Supplier No. 201294100
Add to Cart
Add to Cart
Seahorse 16 row polypropylene reservoir, 4.9ml/row, 54ml max, pyramid base geometries, 19mm height, case of 25