Learn More
Agilent Technologies RES SEAHORSE 24/96 3.5ML 25/CS
Catalog No. 50995877
Encompass
Seahorse unique 24 cavity polypropylene reservoir, 2 rows x 12 columns, 3.5ml/cavity, 84ml max, pyramid base geometries, 19mm height, case of 25
Catalog No. 50-995-877 Supplier Agilent Technologies Supplier No. 201298100
Add to Cart
Add to Cart
Seahorse unique 24 cavity polypropylene reservoir, 2 rows x 12 columns, 3.5ml/cavity, 84ml max, pyramid base geometries, 19mm height, case of 25