Learn More
Agilent Technologies RES SEAHORSE 384/2 155ML 25/CS
Catalog No. 50995878
Encompass
Seahorse unique 5 cavity polypropylene reservoir, 4 control wells (cols 23-24 rows A-H, I-P) of 2.6ml each and 145ml main, 384 pyramid base geometries, 30.6mm height, case of 25
Catalog No. 50-995-878 Supplier Agilent Technologies Supplier No. 201300100
Seahorse unique 5 cavity polypropylene reservoir, 4 control wells (cols 23-24 rows A-H, I-P) of 2.6ml each and 145ml main, 384 pyramid base geometries, 30.6mm height, case of 25