Learn More
Agilent Technologies RES SEAHORSE 384 2C 282ML 25CS
Catalog No. 50995874
Encompass
Seahorse unique 3 cavity polypropylene reservoir, 2 control wells (7ml col 24 rows A-L and 2ml col 24 rows M-P) and 273ml main, 384 pyramid base geometries, 44mm height, case of 25
Catalog No. 50-995-874 Supplier Agilent Technologies Supplier No. 201286100
Seahorse unique 3 cavity polypropylene reservoir, 2 control wells (7ml col 24 rows A-L and 2ml col 24 rows M-P) and 273ml main, 384 pyramid base geometries, 44mm height, case of 25