Learn More
Agilent Technologies RES SEAHORSE 384 W 79ML 25/CS
Supplier:  Agilent Technologies 201278100
Encompass
Seahorse unique 5 cavity polypropylene reservoir, 4 control wells (cols 1,2,11,12) of 3.25ml each and 86.3ml main, 24 pyramid base geometries, 19mm height, case of 25
Catalog No. 50-995-870