Learn More
Agilent Technologies RES SEAHORSE 8 RW 11ML 25/CS
Catalog No. 50995872
Encompass
Seahorse 8 row polypropylene reservoir, 10.6ml/row, 85ml max, pyramid base geometries, 19mm height, case of 25
Catalog No. 50-995-872 Supplier Agilent Technologies Supplier No. 201282100
Add to Cart
Add to Cart
Seahorse 8 row polypropylene reservoir, 10.6ml/row, 85ml max, pyramid base geometries, 19mm height, case of 25