Learn More
Agilent Technologies RES SEAHORSE 8 RW 32ML 25/CS
Catalog No. 50995866
Encompass
Seahorse 8 row polypropylene reservoir, 32ml/row, 256ml max, pyramid base geometries, 44mm height, case of 25
Catalog No. 50-995-866 Supplier Agilent Technologies Supplier No. 201260100
Seahorse 8 row polypropylene reservoir, 32ml/row, 256ml max, pyramid base geometries, 44mm height, case of 25