Learn More
Medchemexpress LLC Resatorvid (TAK-242) | 243984-11-4 | 99.95% | 361.82 | 500 MG
SDP
Catalog No. 5000343163
Encompass_Preferred
Resatorvid (TAK-242) is a selective Toll-like receptor 4 (TLR4) inhibitor. It inhibits NO, TNF-α, and IL-6 production with IC50s of 1.8 nM, 1.9 nM, and 1.3 nM, respectively. This compound downregulates the expression of TLR4 downstream signaling molecules MyD88 and TRIF, inhibits autophagy, and plays a pivotal role in various inflammatory diseases.
  • Selectively inhibits toll-like receptor 4 (TLR4).
  • Inhibits nitric oxide (NO), TNF-α, and IL-6 production.
  • Downregulates the expression of TLR4 downstream signaling molecules MyD88 and TRIF.
  • Inhibits autophagy.
  • Plays a pivotal role in various inflammatory diseases.
  • Suppresses production from LPS-stimulated human peripheral blood mononuclear cells (PBMCs).
  • Reverses LPS-induced body weight loss and muscle loss in mice models.
Catalog No. 50-003-43163 Supplier Medchemexpress LLC Supplier No. HY1110950MG
Resatorvid (TAK-242) is a selective Toll-like receptor 4 (TLR4) inhibitor. It inhibits NO, TNF-α, and IL-6 production with IC50s of 1.8 nM, 1.9 nM, and 1.3 nM, respectively. This compound downregulates the expression of TLR4 downstream signaling molecules MyD88 and TRIF, inhibits autophagy, and plays a pivotal role in various inflammatory diseases.
  • Selectively inhibits toll-like receptor 4 (TLR4).
  • Inhibits nitric oxide (NO), TNF-α, and IL-6 production.
  • Downregulates the expression of TLR4 downstream signaling molecules MyD88 and TRIF.
  • Inhibits autophagy.
  • Plays a pivotal role in various inflammatory diseases.
  • Suppresses production from LPS-stimulated human peripheral blood mononuclear cells (PBMCs).
  • Reverses LPS-induced body weight loss and muscle loss in mice models.