Learn More
Selleck Chemical LLC Resatorvid5mg
SDP
Catalog No. 502173276
Encompass_Preferred
Resatorvid (TAK-242, CLI-095) is a small-molecule-specific inhibitor ofToll-like receptor (TLR) 4 signaling, blocking LPS-induced production of NO, TNF-? and IL-6 in macrophages with IC50 of 1.8 nM, 1.9 nM and 1.3 nM, respectively. Resatorvid downregulates expression of TLR4 downstream signaling molecules MyD88 and TRIF. Resatorvid inhibitsautophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3276 Supplier Selleck Chemical LLC Supplier No. S74555MG
Resatorvid (TAK-242, CLI-095) is a small-molecule-specific inhibitor ofToll-like receptor (TLR) 4 signaling, blocking LPS-induced production of NO, TNF-? and IL-6 in macrophages with IC50 of 1.8 nM, 1.9 nM and 1.3 nM, respectively. Resatorvid downregulates expression of TLR4 downstream signaling molecules MyD88 and TRIF. Resatorvid inhibitsautophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.