Learn More
Biotium Resazurin Cell Viability Assay Kit
Catalog No. NC1192346
Encompass_Preferred
Resazurin Cell Viability Assay offers a rapid, reliable, sensitive, and cost-effective measurement of cell viability. Its performance is comparable to other commercial resazurin-based kits such as alamarBlue, PrestoBlue, or CellTiter-Blue. Cell growth maintains a reducing cellular environment while growth inhibition makes it oxidizing. Resazurin, a redox indicator, measures cell viability by monitoring conversion of oxidized, non-fluorescent resazurin (purple) to the reduced, highly red-fluorescent (blue) resorufin. The fluorescent (Ex/Em 530-560 nm/590 nm) or colorimetric (Abs. 570-600 nm) signal is proportional to the number of live, metabolically active cells. The ready-to-use, single-step, no-wash, no-lysis, homogenous assay can accurately measure as few as 80 cells. Non-toxic reagents allow long-term kinetic monitoring. The assay is non-radioactive, and non-carcinogenic and compatible with a variety of cell types including bacteria, protozoa, yeast, fungi, or mammalian cells.
Catalog No. NC1192346 Supplier Biotium Supplier No. 300252
Add to Cart
Add to Cart
Resazurin Cell Viability Assay offers a rapid, reliable, sensitive, and cost-effective measurement of cell viability. Its performance is comparable to other commercial resazurin-based kits such as alamarBlue, PrestoBlue, or CellTiter-Blue. Cell growth maintains a reducing cellular environment while growth inhibition makes it oxidizing. Resazurin, a redox indicator, measures cell viability by monitoring conversion of oxidized, non-fluorescent resazurin (purple) to the reduced, highly red-fluorescent (blue) resorufin. The fluorescent (Ex/Em 530-560 nm/590 nm) or colorimetric (Abs. 570-600 nm) signal is proportional to the number of live, metabolically active cells. The ready-to-use, single-step, no-wash, no-lysis, homogenous assay can accurately measure as few as 80 cells. Non-toxic reagents allow long-term kinetic monitoring. The assay is non-radioactive, and non-carcinogenic and compatible with a variety of cell types including bacteria, protozoa, yeast, fungi, or mammalian cells.