Learn More
Medchemexpress LLC HY-111391 500mg Medchemexpress, Resazurin (sodium) CAS:62758-13-8 Purity:>98%
SDP
Catalog No. 501968855
Encompass_Preferred
Medchemexpress, HY-111391 500mg Resazurin (sodium) CAS:62758-13-8 Resazurin sodium (Diazoresorcinol sodium) is commonly used to measure bacterial and eukaryotic cell viability through its reduction to the fluorescent product resorufin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8855 Supplier Medchemexpress LLC Supplier No. HY111391500MG
Medchemexpress, HY-111391 500mg Resazurin (sodium) CAS:62758-13-8 Resazurin sodium (Diazoresorcinol sodium) is commonly used to measure bacterial and eukaryotic cell viability through its reduction to the fluorescent product resorufin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.