Learn More
Medchemexpress LLC Resazurin sodium | 62758-13-8 | 98.1% | 251.17 | 1 ML
SDP
Catalog No. 5000343160
Encompass_Preferred
Resazurin sodium (Diazoresorcinol sodium) is a non-toxic, stable, membrane-permeable blue non-fluorescent dye (faintly fluorescent). It functions as a redox indicator, capable of being reduced to the pink, highly fluorescent Resorufin (Ex=530-560 nm, Em=590 nm) in living cells. This compound can be utilized for detecting cell viability, toxicity, proliferation, migration, and invasion in various cell types including human, plant, animal, bacterial, and fungal cells.
  • Functions as a redox indicator
  • Capable of being reduced to highly fluorescent Resorufin in living cells
  • Can be used for detecting cell viability
  • Applicable for toxicity and proliferation studies
  • Suitable for migration and invasion detection
Catalog No. 50-003-43160 Supplier Medchemexpress LLC Supplier No. HY11139110MM/1ML
Resazurin sodium (Diazoresorcinol sodium) is a non-toxic, stable, membrane-permeable blue non-fluorescent dye (faintly fluorescent). It functions as a redox indicator, capable of being reduced to the pink, highly fluorescent Resorufin (Ex=530-560 nm, Em=590 nm) in living cells. This compound can be utilized for detecting cell viability, toxicity, proliferation, migration, and invasion in various cell types including human, plant, animal, bacterial, and fungal cells.
  • Functions as a redox indicator
  • Capable of being reduced to highly fluorescent Resorufin in living cells
  • Can be used for detecting cell viability
  • Applicable for toxicity and proliferation studies
  • Suitable for migration and invasion detection