Learn More
Medchemexpress LLC Resazurin sodium (Diazoresorcinol sodium) | 62758-13-8 | 98.1% | 251.17 | 1 MG
SDP
Catalog No. 5000343806
Encompass_Preferred
Resazurin sodium (Diazoresorcinol sodium) is a non-toxic, stable, membrane-permeable blue non-fluorescent dye. It is used as a redox indicator and can be reduced to pink, highly fluorescent Resorufin (Ex=530-560 nm, Em=590 nm) in living cells. This compound can be used for the detection of cell viability, toxicity, proliferation, migration, and invasion in human, plant, animal, bacterial, and fungal cells, and is commonly employed to measure bacterial and eukaryotic cell viability through its reduction to the fluorescent product resorufin.
  • Non-toxic, stable, and membrane-permeable blue non-fluorescent dye
  • Functions as a redox indicator
  • Reduces to pink, highly fluorescent Resorufin in living cells
  • Used for detection of cell viability, toxicity, proliferation, migration, and invasion
  • Applicable in human, plant, animal, bacterial, and fungal cells
  • Measures bacterial and eukaryotic cell viability
Catalog No. 50-003-43806 Supplier Medchemexpress LLC Supplier No. HY1113915G
Resazurin sodium (Diazoresorcinol sodium) is a non-toxic, stable, membrane-permeable blue non-fluorescent dye. It is used as a redox indicator and can be reduced to pink, highly fluorescent Resorufin (Ex=530-560 nm, Em=590 nm) in living cells. This compound can be used for the detection of cell viability, toxicity, proliferation, migration, and invasion in human, plant, animal, bacterial, and fungal cells, and is commonly employed to measure bacterial and eukaryotic cell viability through its reduction to the fluorescent product resorufin.
  • Non-toxic, stable, and membrane-permeable blue non-fluorescent dye
  • Functions as a redox indicator
  • Reduces to pink, highly fluorescent Resorufin in living cells
  • Used for detection of cell viability, toxicity, proliferation, migration, and invasion
  • Applicable in human, plant, animal, bacterial, and fungal cells
  • Measures bacterial and eukaryotic cell viability