Learn More
Apexbio Technology LLC Resazurin sodium salt 62758-13-8 1g
SDP
Catalog No. 5000092771
Encompass_Preferred
Resazurin sodium salt (62758-13-8) is a cell-permeable fluorometric redox indicator It is designed to assess cellular metabolic activity by measuring the redox potential within viable cells Resazurin sodium salt exerts its biological activity primarily through reduction by intracellular enzymes converting from a non-fluorescent compound to the fluorescent dye resorufin In cell-based studies previously published data demonstrated IC50 values in mammalian cell lines typically in the micromolar range upon prolonged incubation periods Based on these pharmacological properties resazurin sodium salt holds research potential in evaluating cell proliferation metabolic state cytotoxicity and viability assays in high-throughput screening fluorescence microscopy and flow cytometry
Catalog No. 50-000-92771 Supplier Apexbio Technology LLC Supplier No. B60981000
Resazurin sodium salt (62758-13-8) is a cell-permeable fluorometric redox indicator It is designed to assess cellular metabolic activity by measuring the redox potential within viable cells Resazurin sodium salt exerts its biological activity primarily through reduction by intracellular enzymes converting from a non-fluorescent compound to the fluorescent dye resorufin In cell-based studies previously published data demonstrated IC50 values in mammalian cell lines typically in the micromolar range upon prolonged incubation periods Based on these pharmacological properties resazurin sodium salt holds research potential in evaluating cell proliferation metabolic state cytotoxicity and viability assays in high-throughput screening fluorescence microscopy and flow cytometry