Learn More
Abcam Resazurin sodium salt, dye for metabolic activity measurement, 5G
Catalog No. 502705148
Encompass_Preferred
MW 251.17 Da, Purity >80%. Blue non-fluorescent dye and red fluorescent reagent. Redox indicator. Reduces to fluorescent red resorufin (ab145435, ab145372). Cell viability assay reagent.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5148 Supplier Abcam Supplier No. AB1455135G
MW 251.17 Da, Purity >80%. Blue non-fluorescent dye and red fluorescent reagent. Redox indicator. Reduces to fluorescent red resorufin (ab145435, ab145372). Cell viability assay reagent.
The product is subject to the following: Abcam Restricted Use Statement