Learn More
Cayman Chemical Resazurin (sodium salt), Purity: ≥85%, CAS: 62758-13-8, 25 g
SDP
Catalog No. 502952661
Encompass_Preferred
A blue non-fluorescent dye that is used as an oxidation-reduction indicator in cell viability assays in a variety of cells; can be reduced to the highly red-fluorescent product resorufin (ex/em max = 570/580 nm, respectively) and used as a quantifiable detection agent in enzyme activity or gene expression assays
Catalog No. 50-295-2661 Supplier Cayman Chemical Supplier No. 1432225 G
A blue non-fluorescent dye that is used as an oxidation-reduction indicator in cell viability assays in a variety of cells; can be reduced to the highly red-fluorescent product resorufin (ex/em max = 570/580 nm, respectively) and used as a quantifiable detection agent in enzyme activity or gene expression assays