Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rescinnamine 98%, Thermo Scientific™

Catalog No. AC132270010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC132270010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC132270010 Supplier Thermo Scientific Chemicals Supplier No. 132270010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24815-24-5
Molecular Formula C35H42N2O9
Molecular Weight (g/mol) 634.726
InChI Key SZLZWPPUNLXJEA-QEGASFHISA-N
Synonym rescinnamine, rescinnamin, moderil, cinnaloid, rescisan, resealoid, scinnamina, apoterin, anaprel, cartric
PubChem CID 5280954
ChEBI CHEBI:28572
IUPAC Name methyl (1R,15S,17R,18R,19S,20S)-6,18-dimethoxy-17-[(E)-3-(3,4,5-trimethoxyphenyl)prop-2-enoyl]oxy-1,3,11,12,14,15,16,17,18,19,20,21-dodecahydroyohimban-19-carboxylate
SMILES COC1C(CC2CN3CCC4=C(C3CC2C1C(=O)OC)NC5=C4C=CC(=C5)OC)OC(=O)C=CC6=CC(=C(C(=C6)OC)OC)OC

Specifications

Color White to Yellow
Infrared Spectrum Authentic
Quantity 1 g
Merck Index 15, 8262
Formula Weight 634.71
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material Rescinnamine, 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.