Learn More
Description
- White write-on area suitable for pencils, pens, or markers
- Dimension measures 4 mm thick
- Durable resealable plastic evidence bags designed for secure collection and clear documentation.
- Made from strong polyethylene, each bag features a Write-on panel for labeling critical case details and a reliable seal to help maintain chain of custody.
- Ideal for storing and transporting a wide range of forensic evidence.
Specifications
Specifications
| Dimensions (L x W) | 2.5 x 3 in. |
| Product Type | Evidence Bag |
| Color | Transparent with White Label |
| Quantity | 100/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
