Learn More
BIOHIPPO Research Grade Anti-HRSV-A F/Fusion glycoprotein F0 Biosimilar Antibody (191E4#), 100 µg
SDP
Catalog No. 5000858661
Encompass_Preferred
HRSV-A F/Fusion glycoprotein F0 is a research-grade biosimilar antibody. It reacts with HumanrespiratorysyncytialvirusA. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.
Catalog No. 50-008-58661 Supplier BIOHIPPO Supplier No. BHA21415082
HRSV-A F/Fusion glycoprotein F0 is a research-grade biosimilar antibody. It reacts with HumanrespiratorysyncytialvirusA. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.