Learn More
BIOHIPPO Research Grade Anti-Human CD142/F3/TF Biosimilar Antibody (CNTO 860), 100 µg
SDP
Catalog No. 5000859037
Encompass_Preferred
CD142/F3/TF is a research-grade biosimilar antibody. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.
Catalog No. 50-008-59037 Supplier BIOHIPPO Supplier No. BHA21414059
CD142/F3/TF is a research-grade biosimilar antibody. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.