Learn More
BIOHIPPO Research Grade Anti-Human CD205/LY75/DEC-205 Biosimilar Antibody (CDX-1401), 100 µg
SDP
Catalog No. 5000859000
Encompass_Preferred
CDX-1401 is a research-grade biosimilar antibody targeting CD205/LY75/DEC-205. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.
Catalog No. 50-008-59000 Supplier BIOHIPPO Supplier No. BHA21414693
CDX-1401 is a research-grade biosimilar antibody targeting CD205/LY75/DEC-205. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.