Learn More
BIOHIPPO Research Grade Anti-Human CD223/LAG3 Biosimilar Antibody (IMP-701), 100 µg
SDP
Catalog No. 5000859346
Encompass_Preferred
IMP-701 is a research-grade biosimilar antibody targeting CD223/LAG3. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.
Catalog No. 50-008-59346 Supplier BIOHIPPO Supplier No. BHA21414241
IMP-701 is a research-grade biosimilar antibody targeting CD223/LAG3. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.