Learn More
BIOHIPPO Research Grade Anti-Human CD340/ERBB2/HER2/NEU Biosimilar Antibody (ADCT-502), 100 µg
SDP
Catalog No. 5000858574
Encompass_Preferred
ADCT-502 is a research-grade biosimilar antibody targeting CD340/ERBB2/HER2/NEU. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.
Catalog No. 50-008-58574 Supplier BIOHIPPO Supplier No. BHA21414413
Add to Cart
Add to Cart
ADCT-502 is a research-grade biosimilar antibody targeting CD340/ERBB2/HER2/NEU. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.