Learn More
BIOHIPPO Research Grade Anti-Human CD340/ERBB2/HER2/NEU Biosimilar Antibody (CMI Cuba 5G4), 100 µg
SDP
Catalog No. 5000859366
Encompass_Preferred
CD340/ERBB2/HER2/NEU is a research-grade biosimilar antibody. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.
Catalog No. 50-008-59366 Supplier BIOHIPPO Supplier No. BHA21414398
CD340/ERBB2/HER2/NEU is a research-grade biosimilar antibody. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.