Learn More
BIOHIPPO Research Grade Anti-Human DLL3 & CD47/MER6 Biosimilar Antibody (PT-217), 100 µg
SDP
Catalog No. 5000860413
Encompass_Preferred
PT-217 is a research-grade biosimilar antibody targeting DLL3 CD47/MER6. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.
Catalog No. 50-008-60413 Supplier BIOHIPPO Supplier No. BHA21413456
PT-217 is a research-grade biosimilar antibody targeting DLL3 CD47/MER6. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.