Learn More
BIOHIPPO Research Grade Anti-Human EGFR/ERBB1/HER1 Biosimilar Antibody (SYN-004), 100 µg
SDP
Catalog No. 5000858687
Encompass_Preferred
SYN-004 is a research-grade biosimilar antibody targeting EGFR/ERBB1/HER1. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.
Catalog No. 50-008-58687 Supplier BIOHIPPO Supplier No. BHA21414666
SYN-004 is a research-grade biosimilar antibody targeting EGFR/ERBB1/HER1. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.