Learn More
BIOHIPPO Research Grade Anti-Human HGF Biosimilar Antibody (TAK-701), 100 µg
SDP
Catalog No. 5000859371
Encompass_Preferred
TAK-701 is a research-grade biosimilar antibody targeting HGF. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.
Catalog No. 50-008-59371 Supplier BIOHIPPO Supplier No. BHA21414355
TAK-701 is a research-grade biosimilar antibody targeting HGF. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.