Learn More
BIOHIPPO Research Grade Anti-Human IL10 Biosimilar Antibody (MK-1966), 100 µg
SDP
Catalog No. 5000859407
Encompass_Preferred
MK-1966 is a research-grade biosimilar antibody targeting IL10. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.
Catalog No. 50-008-59407 Supplier BIOHIPPO Supplier No. BHA21414331
Add to Cart
Add to Cart
MK-1966 is a research-grade biosimilar antibody targeting IL10. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.