Learn More
BIOHIPPO Research Grade Anti-Human IL12B/IL-12 p40/NKSF2 Biosimilar Antibody (ABT-147), 100 µg
SDP
Catalog No. 5000859935
Encompass_Preferred
ABT-147 is a research-grade biosimilar antibody targeting IL12B/IL-12 p40/NKSF2. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.
Catalog No. 50-008-59935 Supplier BIOHIPPO Supplier No. BHA21413922
ABT-147 is a research-grade biosimilar antibody targeting IL12B/IL-12 p40/NKSF2. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.