Learn More
BIOHIPPO Research Grade Anti-Human MET/c-Met/HGFR Biosimilar Antibody (ARGX-111), 100 µg
SDP
Supplier:  BIOHIPPO BHA21414762
Encompass_Preferred
ARGX-111 is a research-grade biosimilar antibody targeting MET/c-Met/HGFR. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.
Catalog No. 50-008-60275