Learn More
BIOHIPPO Research Grade Anti-Human MET/c-Met/HGFR Biosimilar Antibody (hucMet27-DGN549), 100 µg
SDP
Catalog No. 5000858803
Encompass_Preferred
MET/c-Met/HGFR is a research-grade biosimilar antibody. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.
Catalog No. 50-008-58803 Supplier BIOHIPPO Supplier No. BHA21414760
MET/c-Met/HGFR is a research-grade biosimilar antibody. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.