Learn More
BIOHIPPO Research Grade Anti-Human SLC34A2/NaPi2b Biosimilar Antibody (XMT-1592), 100 µg
SDP
Catalog No. 5000858883
Encompass_Preferred
XMT-1592 is a research-grade biosimilar antibody targeting SLC34A2/NaPi2b. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.
Catalog No. 50-008-58883 Supplier BIOHIPPO Supplier No. BHA21414201
XMT-1592 is a research-grade biosimilar antibody targeting SLC34A2/NaPi2b. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.