Learn More
BIOHIPPO Research Grade Anti-Human TM4SF1/M3S1 Biosimilar Antibody (AGX-A07), 100 µg
SDP
Catalog No. 5000858593
Encompass_Preferred
AGX-A07 is a research-grade biosimilar antibody targeting TM4SF1/M3S1. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.
Catalog No. 50-008-58593 Supplier BIOHIPPO Supplier No. BHA21414118
AGX-A07 is a research-grade biosimilar antibody targeting TM4SF1/M3S1. It reacts with Human. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.