Learn More
BIOHIPPO Research Grade Anti-RSV pre-F/pre-Fusion glycoprotein F0 Biosimilar Antibody (ADI-14442), 100 µg
SDP
Supplier:  BIOHIPPO BHA21414606
Encompass_Preferred
ADI-14442 is a research-grade biosimilar antibody targeting RSV pre-F/pre-Fusion glycoprotein F0. It reacts with HumanrespiratorysyncytialvirusA. Suitable for ELISA, FACS, Functional assay, Research in vivo. This research-grade biosimilar supports comparability, functional, PK/PD and immunogenicity studies. Supplied as 100 ug.
Catalog No. 50-008-58781