Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Research Grade Fetal Bovine Serum, US Sourced

Catalog No. FB12999105 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FB12999105 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FB12999105

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Research Grade Fetal Bovine Serum that is US sourced and triple 0.1 μm filtered.

  • US Origin is compliant with current good manufacturing practices (cGMP).
  • For research use only.
TRUSTED_SUSTAINABILITY

Specifications

Additional Assays Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, Glutamic Pyruvic Transaminase (SGPT), Glutamic Oxaloacetic Transaminase (SGOT), pH, Total Protein (Ph. Eur. 2.5.33), Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality (Ph. Eur. 2.2.35), Iron, Total Iron Binding Capacity (TIBC), Percent Saturation, Glucose, IgG - Nephelometer
Age Fetal
Country of Origin United States
Form Liquid
Format Bottle
Product Type Fetal Bovine Serum
Quantity 500 mL
Serum Treatment Sterile-filtered
Sterility Sterile-filtered
Sterilization Method Triple-filtered, 0.1 μm
Grade Research
Processing Yes
For Use With (Application) For Further Manufacturing Or Research Use. Not For Diagnostic Or Therapeutic Use.
Heat Inactivated No
Hemoglobin Concentration < 35 mg/dL
Lyophilized No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.