Learn More
Pipette Supplies, Inc. Research Pro Battery, NiMH
SDP
Catalog No. NC2060413
Encompass
Research Pro Electronic Single Channel and Multichannel Pipettes Rechargeable Battery Set, NiMH. Eppendorf discontinued theirs; but we still make ours. 1,200mAh. Green 2 cell unit. Our brand replacement batteries are specifically suited for the Research Pro Electronic Pipettes. For Single and Multichannel instruments. Our high quality replacement parts must pass rigorous QC criteria, inspections and performance testing before they are put on our shelves. We guarantee your satisfaction with our replacement parts or your money back. Battery must be charged prior to use. See the instructions included or the user manual below for installation.
Catalog No. NC2060413 Supplier Pipette Supplies, Inc. Supplier No. PS 022467018
Research Pro Electronic Single Channel and Multichannel Pipettes Rechargeable Battery Set, NiMH. Eppendorf discontinued theirs; but we still make ours. 1,200mAh. Green 2 cell unit. Our brand replacement batteries are specifically suited for the Research Pro Electronic Pipettes. For Single and Multichannel instruments. Our high quality replacement parts must pass rigorous QC criteria, inspections and performance testing before they are put on our shelves. We guarantee your satisfaction with our replacement parts or your money back. Battery must be charged prior to use. See the instructions included or the user manual below for installation.