Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Researching Alzheimer's Disease by ELISA Kit
SDPSCI_ED

Catalog No. S27563 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
9 to Undergraduate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S27563 9 to Undergraduate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27563 Supplier Edvotek™ Supplier No. 1116
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Students will become neuroscientists searching for Alzheimer's Disease biomarkers. Explores potential biomarkers by analyzing simulated patient samples from control and Alzheimer's Disease populations.

  • Storage: Some Components Require Refrigerator Storage
  • Completion Time: 1 Class Period
  • For 10 Lab Groups

Specifications

Grade 9 to Undergraduate
Age 14 to 20
Includes 10 x ELISA Wash Buffer, ELISA Dilution Buffer, Aß, Primary Antibody, Secondary Antibody, ABTS, ABTS Reaction Buffer, Stop Solution, Small Transfer Pipettes, Strip Tubes, 15 mL Plastic Tube, 1.5 mL Snap-top Tubes
Requires Paper Towels, Distilled or Deionized Water, Beakers/Flasks, Glassware, Gloves, Safety Goggles, Automatic Micropipettes (recommended)
Time Range 1 Class Period
Content And Storage Some Components Require Refrigerator Storage
Class Size 10 Lab Groups
Product Type Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.