Learn More
Arcticwhite LLC 384 Well Reservoir, Low Profile19mm Height, Pyramid Bottom, 384-well Compatible, Low Profile, Polypropylene (PP), 92 ML, 25 Reagent Reservoirs per Case
SDP
Catalog No. NC3502036
Encompass
384 Well Reservoir, Low Profile, Pyramid Bottom, 384-well Compatible, Low Profile 19mm Height, Polypropylene (PP), 92 ML, 25 Reagent Reservoirs per Case. This reservoir supplies reagents for automated liquid handling and high-throughput screening. Supplied by ArcticWhite as 25 reagent reservoirs per case.
Catalog No. NC3502036 Supplier Arcticwhite LLC Supplier No. AWLSS30022
384 Well Reservoir, Low Profile, Pyramid Bottom, 384-well Compatible, Low Profile 19mm Height, Polypropylene (PP), 92 ML, 25 Reagent Reservoirs per Case. This reservoir supplies reagents for automated liquid handling and high-throughput screening. Supplied by ArcticWhite as 25 reagent reservoirs per case.