Learn More
Apexbio Technology LLC Reserpine 50-55-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062705
Encompass_Preferred
Reserpine a naturally derived alkaloid isolated from plants of the genus Rauvolfia functions primarily through the inhibition of vesicular monoamine transporter 2 (VMAT2) By blocking VMAT2-mediated uptake reserpine causes depletion of monoamine neurotransmitters such as dopamine norepinephrine and serotonin from synaptic storage vesicles In biochemical assays reserpine inhibits VMAT2 activity with an IC50 value typically ranging from 30-50 nM depending on assay specifics In biomedical and pharmacological research this compound is commonly utilized to model neurotransmitter depletion mechanisms in vitro and in animal models facilitating studies on monoamine-related neurological disorders neurotransmission pathways as well as serotonergic dopaminergic and adrenergic signaling
Catalog No. 50-000-62705 Supplier Apexbio Technology LLC Supplier No. N18675.1
Reserpine a naturally derived alkaloid isolated from plants of the genus Rauvolfia functions primarily through the inhibition of vesicular monoamine transporter 2 (VMAT2) By blocking VMAT2-mediated uptake reserpine causes depletion of monoamine neurotransmitters such as dopamine norepinephrine and serotonin from synaptic storage vesicles In biochemical assays reserpine inhibits VMAT2 activity with an IC50 value typically ranging from 30-50 nM depending on assay specifics In biomedical and pharmacological research this compound is commonly utilized to model neurotransmitter depletion mechanisms in vitro and in animal models facilitating studies on monoamine-related neurological disorders neurotransmission pathways as well as serotonergic dopaminergic and adrenergic signaling