Learn More
Medchemexpress LLC Reserpine-d9 | 84759-11-5 | 617.73 | 5 MG
SDP
Catalog No. 5000389831
Encompass_Preferred
Reserpine-d9 is the deuterium labeled Reserpine. Reserpine is an inhibitor of the vesicular monoamine transporter 2 (VMAT2). Stable heavy isotopes have been incorporated into this compound for quantitation during drug development. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Has the potential to affect the pharmacokinetic and metabolic profiles of drugs due to deuteration.
Catalog No. 50-003-89831 Supplier Medchemexpress LLC Supplier No. HYN0480S5MG
Reserpine-d9 is the deuterium labeled Reserpine. Reserpine is an inhibitor of the vesicular monoamine transporter 2 (VMAT2). Stable heavy isotopes have been incorporated into this compound for quantitation during drug development. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Has the potential to affect the pharmacokinetic and metabolic profiles of drugs due to deuteration.